missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloroquinoline-3-carboxylate, 96%, Thermo Scientific™

Product Code. 11409270
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409270 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11409270 Supplier Thermo Scientific Alfa Aesar Supplier No. H50150.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 4-chloroquinoline-3-carboxylate is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13720-94-0
Molecular Formula C12H10ClNO2
Molecular Weight (g/mol) 235.667
MDL Number MFCD00173407
InChI Key DWXQUAHMZWZXHP-UHFFFAOYSA-N
Synonym ethyl 4-chloro-3-quinolinecarboxylate, 4-chloroquinoline-3-carboxylic acid ethyl ester, ethyl 4-chloro-quinoline-3-carboxylate, 3-quinolinecarboxylic acid, 4-chloro-, ethyl ester, pubchem19592, acmc-20ai5d, ethylchloroquinolinecarboxylate, ethyl 4-chloroquinolin-3-carboxylate, ethyl 4-chloro-3-quinoline carboxylate, ethyl 4-chloranylquinoline-3-carboxylate
PubChem CID 268963
IUPAC Name ethyl 4-chloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=C(C2=CC=CC=C2N=C1)Cl

Specifications

Quantity 1 g
Formula Weight 235.67
Percent Purity 96%
Chemical Name or Material Ethyl 4-chloroquinoline-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.