missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-cyanophenylacetate, 98%, Thermo Scientific Chemicals

Product Code. 11302029
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302029 1 g 1g
11312029 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302029 Supplier Thermo Scientific Alfa Aesar Supplier No. H26078.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1528-41-2
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD09953133
InChI Key DFEWKWBIPMKGFG-UHFFFAOYSA-N
Synonym ethyl 2-4-cyanophenyl acetate, ethyl 4-cyanophenylacetate, ethyl 4-cyanophenyl acetate, 4-cyano-phenyl acetic acid ethyl ester, ethyl p-cyanophenylacetate, acmc-1bve3, ethyl 4-cyanobenzeneacetate, ethyl4-cyanophenylacetate, 4-cyanophenylacetic acid ethyl ester
PubChem CID 279718
IUPAC Name ethyl 2-(4-cyanophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)C#N

Specifications

Melting Point 90°C to 94°C
Quantity 1 g
UN Number UN3439
Formula Weight 189.21
Percent Purity 98%
Chemical Name or Material Ethyl 4-cyanophenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.