missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide(Piroxicam Impurity H), TRC

Product Code. 30375356
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375356 100 mg -
30373470 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375356 Supplier Toronto Research Chemicals Supplier No. TRCE918850100MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24683-21-4
Molecular Formula C11 H11 N O5 S
Molecular Weight (g/mol) 269.27
Synonym Ethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide, Piroxicam Imp. H (EP)
IUPAC Name ethyl 4-hydroxy-1,1-dioxo-2H-1λ^{6},2-benzothiazine-3-carboxylate
SMILES CCOC(=O)C1=C(O)c2ccccc2S(=O)(=O)N1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H11NO5S/c1-2-17-11(14)9-10(13)7-5-3-4-6-8(7)18(15,16)12-9/h3-6,12-13H,2H2,1H3
Formula Weight 269.0358
Chemical Name or Material Ethyl 4-Hydroxy-2H-1,2-benzothiazine-3-carboxylate 1,1-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.