missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxy-3-methoxycinnamate, 98%

Product Code. 11318096
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318096 250 mg 250mg
11308096 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318096 Supplier Thermo Scientific Alfa Aesar Supplier No. L14960.MD
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4046-02-0 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4046-02-0
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009190
InChI Key ATJVZXXHKSYELS-FNORWQNLSA-N
Synonym ethyl ferulate, ferulic acid ethyl ester, ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, unii-5b8915uelw, ethyl 4'-hydroxy-3'-methoxycinnamate, e-ethyl 3-4-hydroxy-3-methoxyphenyl acrylate, ethyl 2e-3-4-hydroxy-3-methoxyphenyl prop-2-enoate, ethyl 4-hydroxy-3-methoxycinnamate, e-3-4-hydroxy-3-methoxy-phenyl-acrylic acid ethyl ester, ethyl 4-hydroxy-3-methoxycinnamate ferulic acid ethyl ester
PubChem CID 736681
IUPAC Name ethyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 56°C to 58°C
Quantity 250 mg
Beilstein 2696807
Sensitivity Light sensitive
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Ethyl 4-hydroxy-3-methoxycinnamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.