missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%

Product Code. 11471080
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471080 25 g 25g
11461080 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471080 Supplier Thermo Scientific Alfa Aesar Supplier No. B25572.14
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17159-80-7 | C9H16O3 | 172.22 g/mol

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17159-80-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00192156,MFCD16295187
InChI Key BZKQJSLASWRDNE-UHFFFAOYSA-N
Synonym ethyl 4-hydroxycyclohexanecarboxylate, trans-ethyl 4-hydroxycyclohexanecarboxylate, ethyl trans-4-hydroxycyclohexanecarboxylate, 4-hydroxy-cyclohexanecarboxylic acid ethyl ester, ethyl 4-hydroxycyclohexane carboxylate, cis-ethyl 4-hydroxycyclohexanecarboxylate, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, 4-hydroxycyclohexanecarboxylic acid ethyl ester, trans-4-hydroxycyclohexanecarboxylic acid ethyl ester, cyclohexanecarboxylic acid, 4-hydroxy-, ethyl ester, cis
PubChem CID 86973
IUPAC Name ethyl 4-hydroxycyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(O)CC1

Specifications

Density 1.068
Boiling Point 127°C to 134°C (0.1 mmHg)
Flash Point >110(230°F)
Refractive Index 1.467
Quantity 25 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.