missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-nitroindole-2-carboxylate, 97%

Product Code. 15440139
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440139 250 mg 250mg
15430139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440139 Supplier Thermo Scientific Alfa Aesar Supplier No. H63440.MD
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4993-93-5 | C11H10N2O4 | 234.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4993-93-5
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.211
MDL Number MFCD06795512
InChI Key LXUABEANNNKOLF-UHFFFAOYSA-N
Synonym ethyl 4-nitroindole-2-carboxylate, 4-nitro-indole-2-carboxylic acid ethyl ester, 4-nitro-1h-indole-2-carboxylic acid ethyl ester, ethyl-4-nitroindole-2-carboxylate, ethyl 4-nitro-2-indolecarboxylate, 1h-indole-2-carboxylic acid, 4-nitro-, ethyl ester
PubChem CID 283173
IUPAC Name ethyl 4-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC=C2[N+](=O)[O-]

Specifications

Color Yellow
Quantity 250 mg
Formula Weight 234.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 4-nitroindole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.