missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-quinazolone-2-carboxylate, 98%

Product Code. 11386966
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11386966

Brand: Thermo Scientific Alfa Aesar L06709.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29113-33-5 | C11H10N2O3 | 218.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29113-33-5
Molecular Formula C11H10N2O3
Molecular Weight (g/mol) 218.212
MDL Number MFCD00051583
InChI Key BMCAWNQKVVTNFP-UHFFFAOYSA-N
Synonym ethyl 4-quinazolone-2-carboxylate, ethyl 4-oxo-3,4-dihydroquinazoline-2-carboxylate, ethyl 4-hydroxyquinazoline-2-carboxylate, ethyl 4-oxo-3,4-dihydro-2-quinazolinecarboxylate, ethyl 4-oxo-3h-quinazoline-2-carboxylate, 4-quinazolone-2-carboxylic acid ethyl ester, 4-hydroxy-quinazoline-2-carboxylic acid ethyl ester, 2-quinazolinecarboxylic acid, 1,4-dihydro-4-oxo-, ethyl ester, acmc-20aic8, enamine_001988
PubChem CID 293295
IUPAC Name ethyl 4-oxo-1H-quinazoline-2-carboxylate
SMILES CCOC(=O)C1=NC(=O)C2=CC=CC=C2N1

Specifications

Melting Point 189°C to 193°C
Quantity 1 g
Beilstein 190186
Formula Weight 218.21
Percent Purity 98%
Chemical Name or Material Ethyl 4-quinazolone-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.