missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(trifluoromethyl)pyrimidine-5-carboxylate, Thermo Scientific™

Product Code. 10301420
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301420 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10301420 Supplier Thermo Scientific Maybridge Supplier No. MAY00181DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 187035-81-0
Packaging Amber Glass Bottle
Molecular Formula C8H7F3N2O2
Quantity 1 g
Synonym ethyl 4-trifluoromethyl pyrimidine-5-carboxylate, 4-trifluoromethylpyrimidine-5-carboxylic acid ethyl ester, ethyl-4-trifluoromethyl pyrimidine-5-carboxylate
InChI Key MXCQFZJBUCRYHU-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CN=CN=C1C(F)(F)F
IUPAC Name ethyl 4-(trifluoromethyl)pyrimidine-5-carboxylate
Molecular Weight (g/mol) 220.151
PubChem CID 22744070
Formula Weight 220.15
Percent Purity 95%
Chemical Name or Material Ethyl 4-(trifluoromethyl)pyrimidine-5-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.