missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride 0.5 hydrate, 97%, Thermo Scientific™

Product Code. 10216613
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10216613 1 g 1g
10532754 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216613 Supplier Thermo Scientific Maybridge Supplier No. CC53973DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 623564-18-1
Molecular Formula C9H15Cl2N3O2
Molecular Weight (g/mol) 268.14
MDL Number MFCD17015229
InChI Key UVYMXIAFTVXRIT-UHFFFAOYSA-N
Synonym ethyl 5h,6h,7h,8h-imidazo 1,2-a pyrazine-2-carboxylate dihydrochloride, ethyl 5,6,7,8-tetrahydroimidazo 1,2-a pyrazine-2-carboxylate-hydrogen chloride 1/2, ethyl 5,6,7,8-tetrahydroimidazo 1,2-a pyrazine-2-carboxylate dihydrochloride 0.5 hydrate
PubChem CID 24229538
IUPAC Name ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate;dihydrochloride
SMILES Cl.Cl.CCOC(=O)C1=CN2CCNCC2=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 277.15
Percent Purity 97%
Chemical Name or Material Ethyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine-2-carboxylate dihydrochloride 0.5 hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.