missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-1-methylpyrazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 11376865
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Amber Glass Bottle
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11376865 100 g Amber Glass Bottle 100g
10289562 10 g Amber Glass Bottle 10g
10299562 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11376865 Supplier Thermo Scientific Maybridge Supplier No. EN00291FA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 31037-02-2
Molecular Formula C7H11N3O2
Molecular Weight (g/mol) 169.18
MDL Number MFCD00051652
InChI Key MEUSJJFWVKBUFP-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-methyl-1h-pyrazole-4-carboxylate, 5-amino-1-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl5-amino-1-methylpyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid, 5-amino-1-methyl-, ethyl ester, 5-amino-1-methylpyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-1-methyl-4-pyrazolecarboxylate, ethyl 5-amino-1-methyl-pyrazole-4-carboxylate, acmc-1agpk, maybridge1_003833, ethyl-5-amino-1-methyl-1h-pyrazol-4-carboxylat
PubChem CID 271299
SMILES CCOC(=O)C1=C(N)N(C)N=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 100 g
IUPAC Name ethyl 5-amino-1-methylpyrazole-4-carboxylate
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.