missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate, 97+%

Product Code. 11342029
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342029 25 g 25g
11322029 1 g 1g
11332029 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342029 Supplier Thermo Scientific Alfa Aesar Supplier No. H26079.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23286-70-6 | C7H11N3O2 | 169.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23286-70-6
Molecular Formula C7H11N3O2
Molecular Weight (g/mol) 169.184
MDL Number MFCD01210846
InChI Key WOCMIZZYXHVSPS-UHFFFAOYSA-N
Synonym ethyl 5-amino-3-methyl-1h-pyrazole-4-carboxylate, 5-amino-3-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-3-methylpyrazole-4-carboxylate, ethyl 3-amino-5-methyl-2h-pyrazole-4-carboxylate, acmc-1co96, 5-amino-3-methyl-1h-pyrazole-4-carboxylicacidethylester, ethyl 3-amino-5-methylpyrazole-4-carboxylate, 3-amino-5-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-methyl-5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, pyrazole-4-carboxylic acid, 3-amino-5-methyl-, ethyl ester
PubChem CID 683559
IUPAC Name ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1N)C

Specifications

Melting Point 108°C
Quantity 25 g
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.