missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-aminovalerate hydrochloride, 96%

Product Code. 15498137
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498137 1 g 1g
15408147 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498137 Supplier Thermo Scientific Alfa Aesar Supplier No. H51682.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29840-57-1 | C7H16ClNO2 | 181.66 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29840-57-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD12546205
InChI Key JELXJZASXLHKDV-UHFFFAOYSA-N
Synonym ethyl 5-aminovalerate hydrochloride, ethyl 5-aminopentanoate hydrochloride, ethyl 5-aminovalerate hcl, 5-aminovaleric acid ethyl ester hydrochloride, pentanoic acid, 5-amino-, ethyl ester, hydrochloride, ethyl 5-aminopentanoate hcl, ethyl5-aminovaleratehydrochloride, ethyl 5-azanylpentanoate hydrochloride, 5-amino-valeric acid ethyl ester hydrochloride, 5-aminopentanoic acid ethyl ester hydrochloride
PubChem CID 16094861
IUPAC Name ethyl 5-aminopentanoate;hydrochloride
SMILES CCOC(=O)CCCCN.Cl

Specifications

Linear Formula H2N(CH2)4CO2CH2CH3·HCl
Quantity 1 g
Formula Weight 181.66
Percent Purity 96%
Chemical Name or Material Ethyl 5-aminovalerate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.