missing translation for 'onlineSavingsMsg'
Learn More

Ethyl(5-bromobenzofuran)-2-carboxylate, 95%, Thermo Scientific™

Product Code. 10412453
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10412453 250 mg 250mg
10188082 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412453 Supplier Thermo Scientific Maybridge Supplier No. CC31223CB
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 84102-69-2
Molecular Formula C11H9BrO3
Molecular Weight (g/mol) 269.094
InChI Key XLJWAHXKBCDQNP-UHFFFAOYSA-N
PubChem CID 735184
IUPAC Name ethyl 5-bromo-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(O1)C=CC(=C2)Br

Specifications

Assay Percent Range 95%
Quantity 250 mg
Synonym ethyl 5-bromobenzofuran-2-carboxylate, buttpark 34\07-75, 5-bromo-benzofuran-2-carboxylic acid ethyl ester, ethylbromobenzofurancarboxylate, ethyl5-bromobenzofuran-2-carboxylate, ethyl 5-bromo-2-benzofurancarboxylate, ethyl 5-bromo-benzofuran-2-carboxylate, 5-bromo-2-ethoxycarbonyl-1-benzofuran
Formula Weight 269.09
Percent Purity 95%
Chemical Name or Material Ethyl (5-bromobenzofuran)-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.