missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromovalerate, 99%

Product Code. 10347760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10347760 25 mL Glass bottle 25mL
10061680 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347760 Supplier Thermo Scientific Acros Supplier No. 156540250
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14660-52-7 | C7H13BrO2 | 209.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14660-52-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00000266
InChI Key AFRWBGJRWRHQOV-UHFFFAOYSA-N
Synonym ethyl 5-bromovalerate, 5-bromopentanoic acid ethyl ester, pentanoic acid, 5-bromo-, ethyl ester, 5-bromovaleric acid ethyl ester, ethyl omega-bromovalerate, ethyl5-bromovalerate, ethyl-5-bromovalerate, pubchem3955, ethyl-5-bromo-valerate, ethyl-5-bromopentanoate
PubChem CID 84580
IUPAC Name ethyl 5-bromopentanoate
SMILES CCOC(=O)CCCCBr

Specifications

Color Colorless to Yellow
Density 1.3200g/mL
Boiling Point 104°C to 109°C (12.0 mmHg)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)4CO2C2H5
Refractive Index 1.4570 to 1.46
Quantity 25 mL
Beilstein 02, II, 269
Specific Gravity 1.32
Formula Weight 209.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl 5-bromovalerate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 5-bromovalerate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.