missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-chloro-2-indolecarboxylate, 97%, Thermo Scientific™

Product Code. 10216020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10216020 5 g Glass bottle 5g
10551411 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10216020 Supplier Thermo Scientific Acros Supplier No. 160290050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4792-67-0
Molecular Formula C11H10ClNO2
Molecular Weight (g/mol) 223.66
MDL Number MFCD00005610
InChI Key LWKIFKYHCJAIAB-UHFFFAOYSA-N
Synonym ethyl 5-chloroindole-2-carboxylate, ethyl 5-chloro-2-indolecarboxylate, 5-chloroindole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-chloro-, ethyl ester, ethyl-5-chloroindole-2-carboxylate, 5-chloro-1h-indole-2-carboxylic acid ethyl ester, pubchem7173, 5-chloroindole-2-carboxylicacidethylester, acmc-1aoif, 5-chloro-2-indolecarboxylic acid ethyl ester
PubChem CID 78518
IUPAC Name ethyl 5-chloro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)Cl

Specifications

Melting Point 166°C to 171°C
Color Brown or Tan to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 223.66
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material Ethyl 5-chloro-2-indolecarboxylate, 97%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.