missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(chloromethyl)-2-furancarboxylate, 95%

Product Code. 10586831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10586831 1 g Glass bottle 1g
10461972 5 g Glass bottle 5g
10259323 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10586831 Supplier Thermo Scientific Acros Supplier No. 349730010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2528-00-9 | C8H9ClO3 | 188.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2528-00-9
Molecular Formula C8H9ClO3
Molecular Weight (g/mol) 188.61
MDL Number MFCD00173837
InChI Key JBACYJRMCXLIQU-UHFFFAOYSA-N
Synonym ethyl 5-chloromethyl-2-furoate, ethyl 5-chloromethyl furan-2-carboxylate, ethyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-2-furancarboxylic acid ethyl ester, 2-furancarboxylic acid, 5-chloromethyl-, ethyl ester, buttpark 9650-29, acmc-1cej4, ethyl 2-chloromethyl-5-furoate
PubChem CID 263166
IUPAC Name ethyl 5-(chloromethyl)furan-2-carboxylate
SMILES CCOC(=O)C1=CC=C(O1)CCl

Specifications

Color Yellow
Density 1.2250g/mL
Boiling Point 115.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130
Quantity 1 g
Specific Gravity 1.225
Formula Weight 188.61
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 5-(chloromethyl)-2-furancarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 5-(chloromethyl)-2-furancarboxylate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.