missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-(hydroxymethyl)isoxazole-3-carboxylate, 90%, Thermo Scientific™

Product Code. 10376614
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10376614 5 g Amber Glass Bottle 5g
10374963 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376614 Supplier Thermo Scientific Maybridge Supplier No. MO08038DE
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 123770-62-7
Molecular Formula C7H9NO4
Molecular Weight (g/mol) 171.152
MDL Number MFCD08273501
InChI Key ZWVVVEYXDQMHGQ-UHFFFAOYSA-N
Synonym ethyl 5-hydroxymethyl isoxazole-3-carboxylate, ethyl 5-hydroxymethyl-1,2-oxazole-3-carboxylate, 5-hydroxymethylisoxazole-3-carboxylic acid ethyl ester, 3-isoxazolecarboxylicacid, 5-hydroxymethyl-, ethyl ester, 3-isoxazolecarboxylic acid, 5-hydroxymethyl-, ethyl ester, acmc-1c7ap, ethyl 5-hydroxymethylisoxazole-3-carboxylate, ethyl 5 hydroxymethyl isoxazole-3-carboxylate, ethyl 5-hydroxymethyl-isoxazole-3-carboxylate
PubChem CID 8027233
IUPAC Name ethyl 5-(hydroxymethyl)-1,2-oxazole-3-carboxylate
SMILES CCOC(=O)C1=NOC(=C1)CO

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 171.15
Percent Purity 90%
Chemical Name or Material Ethyl 5-(hydroxymethyl)isoxazole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.