missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromo-2-methylquinoline-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10381353
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10381353

Brand: Thermo Scientific Maybridge BTB00099EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 62482-30-8
Molecular Formula C13H12BrNO2
Molecular Weight (g/mol) 294.15
MDL Number MFCD00115248
InChI Key KOVIYCANWGLBHA-UHFFFAOYSA-N
Synonym maybridge1_000196, 2-methyl-6-bromoquinoline-4-carboxylic acid ethyl ester
PubChem CID 305135
IUPAC Name ethyl 6-bromo-2-methylquinoline-4-carboxylate
SMILES CCOC(=O)C1=C2C=C(Br)C=CC2=NC(C)=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 294.15
Percent Purity 95%
Chemical Name or Material Ethyl 6-bromo-2-methylquinoline-4-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.