missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate, 98%

Product Code. 15428735
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428735 5 g 5g
15438735 250 mg 250mg
15418735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428735 Supplier Thermo Scientific Alfa Aesar Supplier No. H33992.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1027511-41-6 | C9H8BrN3O2 | 270.086 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1027511-41-6
Molecular Formula C9H8BrN3O2
Molecular Weight (g/mol) 270.086
MDL Number MFCD10574908
InChI Key PZZTXIWJXOMKEU-UHFFFAOYSA-N
Synonym ethyl 6-bromopyrazolo 1,5-a pyrimidine-3-carboxylate, 6-bromo-3-ethoxycarbonyl pyrazolo 1,5-a pyrimidine, 6-bromopyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester, pubchem20934, ethyl 6-bromopyrazolo 1,5-a pyrimidine-3-carboxyla, ethyl 6-bromo-pyrazolo 1,5-a pyrimidine-3-carboxylate, ethyl 6-bromopyrazolo 1,5-alpha pyrimidine-3-carboxylate, 6-bromo-pyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester
PubChem CID 45073325
IUPAC Name ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
SMILES CCOC(=O)C1=C2N=CC(=CN2N=C1)Br

Specifications

Melting Point 144°C to 146°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 270.08
Percent Purity 98%
Chemical Name or Material Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.