missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-(chloroformyl)hexanoate, 95%, Thermo Scientific™

Product Code. 11494620
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11494620

Brand: Thermo Scientific Alfa Aesar H50714.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 6-(chloroformyl)hexanoate is used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14794-32-2
Molecular Formula C9H15ClO3
Molecular Weight (g/mol) 206.666
MDL Number MFCD00671507
InChI Key NYZNYLOIPHDFKS-UHFFFAOYSA-N
Synonym ethyl 6-chloroformyl hexanoate, 6-chloroformyl hexanoic acid ethyl ester, heptanoic acid,7-chloro-7-oxo-, ethyl ester, acmc-209czj, ethyl 6-chloroformylhexanoate, ethyl 6-chloroformyl-hexanoate, 6-chlorocarbonylhexanoic acid ethyl ester, heptanoic acid, 7-chloro-7-oxo, ethyl ester
PubChem CID 543232
IUPAC Name ethyl 7-chloro-7-oxoheptanoate
SMILES CCOC(=O)CCCCCC(=O)Cl

Specifications

Linear Formula ClCO-CH2CH2CH2CH2CH2CO2CH2CH3
Quantity 1 g
Formula Weight 206.67
Percent Purity 95%
Chemical Name or Material Ethyl 6-(chloroformyl)hexanoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.