missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-nitroindole-2-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15400329
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400329 1 g 1g
15410329 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400329 Supplier Thermo Scientific Alfa Aesar Supplier No. H63569.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16792-45-3 | C11H10N2O4 | 234.21 g/mol

Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16792-45-3
Molecular Formula C11H10N2O4
Molecular Weight (g/mol) 234.21
MDL Number MFCD04973967
InChI Key SOMZLSPJSSDTAP-UHFFFAOYSA-N
Synonym ethyl 6-nitroindole-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 6-nitro-2-carbethoxyindole, 6-nitro-2-carboethoxyindole, ethyl 6-nitroindol-2-carboxylate, 1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester, 1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester
PubChem CID 616444
IUPAC Name ethyl 6-nitro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O

Especificaciones

Color Yellow
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 234.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 6-nitroindole-2-carboxylate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.