missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-methoxybenzofuran-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10202372 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10202372 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10202372 Supplier Thermo Scientific Acros Supplier No. 356370010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 50551-58-1
Melting Point 85°C to 90°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C12H12O4
MDL Number MFCD00060510
Quantity 1 g
Synonym ethyl 7-methoxybenzofuran-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acid ethyl ester, 7-methoxybenzofuran-2-carboxylic acid, ethyl ester, ethyl 7-methoxybenzo b furan-2-carboxylate, 7-methoxybenzofuran-2-carboxylic acidethyl ester, ethyl7-methoxybenzofuran-2-carboxylate, acmc-1aowz, 2-ethoxycarbonyl-7-methoxybenzofuran, ethyl 4-methoxylbenzofunan-2-carboxylate, ethyl 7-methoxybenzo d furan-2-carboxylate
InChI Key SXCMZYVXJUJIDJ-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC2=CC=CC(OC)=C2O1
IUPAC Name ethyl 7-methoxy-1-benzofuran-2-carboxylate
Molecular Weight (g/mol) 220.22
PubChem CID 521284
Formula Weight 220.23
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material Ethyl 7-methoxybenzofuran-2-carboxylate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.