missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, 99.6%, ACS reagent, meets the requirements of Reag. Ph.Eur.

Product Code. 10607571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum
Unit Size:
1 L
2.5 L
25 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607571 500 mL EcoSafPak™ Glass Bottle 500mL
10589282 1 L Glass Bottle 1 L
10052593 2.5 L Glass Bottle 2.5 L
10247573 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10607571 Supplier Thermo Scientific Acros Supplier No. 423685000
  • ONLINE EXCLUSIVE PRICE
    32.45 €
    31.50 € / 500mL
    Save 0.95 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Density 0.9020g/mL
Quantity 500 mL
Flash Point −4°C
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3710 to 1.3740
Residue after Evaporation 0.003% max.
Assay Percent Range 99.6%
Specific Gravity 0.902
Titratable Acid 0.0009 meq/g max.
Linear Formula CH3CO2C2H5
Viscosity 0.45 mPa.s (20°C)
Water 0.2% max. (K.F.)
Beilstein 02, 125
Merck Index 15, 3811
Boiling Point 75.0°C to 78.0°C
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in benzene, acetone, chloroform, alcohol,, ether
Formula Weight 88.11
Percent Purity 99.6%
Grade ACS
Chemical Name or Material Ethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.