missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl acetate, 99+%, extra pure
Tienda Thermo Scientific Chemicals Productos
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
missing translation for 'unitSize'
1 L
10 L
2.5 L
25 L
5 L
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Melting Point | -83.5°C |
| Density | 0.9020g/mL |
| Boiling Point | 75.0°C to 78.0°C |
| Quantity | 10 L |
| Flash Point | −4°C |
| Packaging | Plastic Drum |
| Physical Form | Liquid |
| Refractive Index | 1.3710 to 1.3740 |
| Assay Percent Range | 99+% |
| Residue after Evaporation | 0.005% max. |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Ethyl acetate
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Ethyl acetate
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido