missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, ACS, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15638450
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 15638450

Brand: Honeywell-Riedel-de Haen 6768104L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.902g/mL at 25°C
Boiling Point 77°C
Quantity 4 L
Explosion Limit 2.2 to 11.5%
Ignition Point 801°C
Packaging Aluminum Bottle
Physical Form Liquid
Linear Formula CH3COOC2H5
Vapor Pressure 100mmHg (20°C)
UN Number UN1173
Beilstein 506104
Solubility Information 80g/L in water at 20°C
Formula Weight 88.11g/mol
Percent Purity ≥99.5%
Chemical Name or Material Ethyl acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.