missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Ethyl Acetate CHROMASOLV™

Product Code. 15965440
Change view
Click to view available options
Quantity:
7 L
18 L
45 L
Unit Size:
18 L
45 L
7 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15965440 7 L 7 L
15945440 18 L 18 L
15955440 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15965440 Supplier Riedel-de Haën Supplier No. 310637L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Ethyl Acetate CHROMASOLV™ for pesticide residue analysis, ≥99.9%.

  • For gas chromatography (GC).
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC.
  • Non-volatile matter and water content are specified for each solvent.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym EtOAc
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

pH No data available
Melting Point -84°C
CAS Min % ≥90.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.901 g/cm3
Boiling Point 77°C (1.013 hPa)
Quantity 7 L
Flash Point 11.5 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV
Linear Formula CH3COOC2H5
Vapor Pressure 97 hPa (20°C)
UN Number UN1173
Viscosity 0.45 mPa.s (20°C)
Concentration ≥99.90%
Percent Purity ≥99.90%
Grade GC
Chemical Name or Material Ethyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.