missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate (Sequencing), Fisher BioReagents

Product Code. 10163333
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163333 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163333 Supplier Fisher BioReagents Supplier No. BP11251
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This solvent is commonly used in DNA or peptide synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -83.5°C
Absorbance 1 max. at 255nm, 0.15 max. at 260nm, 0.025 max. at 270nm
Color Colorless
Density 0.902g/cm³
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3680 to 1.3710 (at 25°C)
Residue after Evaporation 10ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.0009mEq/g max.
Linear Formula CH3COOCH2CH3
Vapor Pressure 103mbar at 20°C
Viscosity 0.45 cP at 20°C
UN Number 1173
Water 0.02% max.
Merck Index 15, 3811
Boiling Point 75°C
Formula Weight 88.11
Percent Purity ≥99.9%
Grade Sequencing
Chemical Name or Material Ethyl Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.