missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetimidate hydrochloride, 97%

Product Code. 11438547
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438547 10 g 10g
11448547 50 g 50g
11458547 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438547 Supplier Thermo Scientific Alfa Aesar Supplier No. A18618.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2208-07-3 | C4H10ClNO | 123.58 g/mol

Ethyl acetimidate hydrochloride was used in preparation of amidinated carbonic anhydrase via chemical modification of human erythrocyte carbonic anhydrase. It was also used in synthesis of methyl 2-methyl-2-thiazoline-4-carboxylate hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acetimidate hydrochloride was used in preparation of amidinated carbonic anhydrase via chemical modification of human erythrocyte carbonic anhydrase. It was also used in synthesis of methyl 2-methyl-2-thiazoline-4-carboxylate hydrochloride.

Solubility
Soluble in water (50mg/L).

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 2208-07-3
MDL Number MFCD00012572

Specifica

Melting Point ∼108°C (decomposition)
Molecular Formula C4H10ClNO
Quantity 10 g
Beilstein 3552401
Sensitivity Moisture sensitive
Synonym ethyl acetimidate hydrochloride, ethyl ethanimidate hydrochloride, ethanimidic acid, ethyl ester, hydrochloride, ethyl ethanimidoate hydrochloride, ethylacetimidatehydrochloride, ethyl iminoacetate hydrochloride, ea hydrochloride, acetimidic acid, hydrochloride, ethanimidic acid, ethyl ester, hydrochloride 1:1, ethanimidic acid, hydrochloride
Solubility Information Soluble in water (50mg/L).
InChI Key WGMHMVLZFAJNOT-UHFFFAOYSA-N
SMILES CCOC(=N)C.Cl
IUPAC Name ethyl ethanimidate;hydrochloride
Molecular Weight (g/mol) 123.58
PubChem CID 2724290
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material Ethyl acetimidate hydrochloride
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.