missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetimidate hydrochloride, 97%

Product Code. 11458547
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458547 250 g 250g
11438547 10 g 10g
11448547 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458547 Supplier Thermo Scientific Alfa Aesar Supplier No. A18618.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2208-07-3 | C4H10ClNO | 123.58 g/mol

Ethyl acetimidate hydrochloride was used in preparation of amidinated carbonic anhydrase via chemical modification of human erythrocyte carbonic anhydrase. It was also used in synthesis of methyl 2-methyl-2-thiazoline-4-carboxylate hydrochloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acetimidate hydrochloride was used in preparation of amidinated carbonic anhydrase via chemical modification of human erythrocyte carbonic anhydrase. It was also used in synthesis of methyl 2-methyl-2-thiazoline-4-carboxylate hydrochloride.

Solubility
Soluble in water (50mg/L).

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2208-07-3
MDL Number MFCD00012572

Specifications

Melting Point ∼108°C (decomposition)
Molecular Formula C12H9NO3
Quantity 250 g
Beilstein 3552401
Sensitivity Moisture sensitive
Synonym 2-nitrodiphenyl ether, 2-nitrophenyl phenyl ether, benzene, 1-nitro-2-phenoxy, o-nitrophenyl phenyl ether, 2-nde, phenyl o-nitrophenyl ether, ether, o-nitrophenyl phenyl, ether, 2-nitrophenyl phenyl, ether, 2-nitrodiphenyl,, ganoderic-acid-g
Solubility Information Soluble in water (50mg/L).
InChI Key VNHGETRQQSYUGZ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2[N+](=O)[O-]
IUPAC Name 1-nitro-2-phenoxybenzene
Molecular Weight (g/mol) 215.21
PubChem CID 16661
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material Ethyl acetimidate hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.