missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetimidate hydrochloride, 97%

Product Code. 10744163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10744163 50 g Glass bottle 50g
10272602 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10744163 Supplier Thermo Scientific Acros Supplier No. 224100500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2208-07-3 | C4H9NO·HCl | 123.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2208-07-3
Molecular Formula C4H9NO·HCl
Molecular Weight (g/mol) 123.58
MDL Number MFCD00012572
InChI Key WGMHMVLZFAJNOT-UHFFFAOYSA-N
Synonym ethyl acetimidate hydrochloride, ethyl ethanimidate hydrochloride, ethanimidic acid, ethyl ester, hydrochloride, ethyl ethanimidoate hydrochloride, ethylacetimidatehydrochloride, ethyl iminoacetate hydrochloride, ea hydrochloride, acetimidic acid, hydrochloride, ethanimidic acid, ethyl ester, hydrochloride 1:1, ethanimidic acid, hydrochloride
PubChem CID 2724290
IUPAC Name ethyl ethanimidate;hydrochloride
SMILES CCOC(=N)C.Cl

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3C(=NH)OC2H5·HCl
Quantity 50 g
Beilstein 02, 182
Formula Weight 123.58
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl acetimidate hydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl acetimidate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.