missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha,alpha-difluorophenylacetate, 99%, Thermo Scientific™

Product Code. 11301959
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301959 1 g 1g
11311959 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301959 Supplier Thermo Scientific Alfa Aesar Supplier No. H26029.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2248-46-6
Molecular Formula C10H10F2O2
Molecular Weight (g/mol) 200.185
MDL Number MFCD08275296
InChI Key KCMSDCHUELUJPX-UHFFFAOYSA-N
Synonym ethyl2,2-difluoro-2-phenylacetate, ethyl difluoro phenyl acetate, alpha,alpha-difluoro phenylacetic acid ethyl ester, difluoro-phenyl-acetic acid ethyl ester, ethyl 2,2-difluoro-2-phenyl-acetate, acmc-1cmi4, difluorophenylacetic acid ethyl ester, ethyl 2,2-difluoro-2-phenylethanoate, ethyl alpha,alpha-difluorophenylacetate, difluoro phenyl acetic acid ethyl ester
PubChem CID 10631860
IUPAC Name ethyl 2,2-difluoro-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)(F)F

Specifications

Refractive Index 1.46
Quantity 1 g
Formula Weight 200.19
Percent Purity 99%
Chemical Name or Material Ethyl alpha,alpha-difluorophenylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.