missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha-bromophenylacetate, 97%

Product Code. 11424587
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424587 10 g 10g
11434587 50 g 50g
11444587 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424587 Supplier Thermo Scientific Alfa Aesar Supplier No. B21114.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2882-19-1 | C10H11BrO2 | 243.10 g/mol

α-Bromobenzeneacetic acid ethyl ester is used as an alkyl halide initiator in controlled/living radical polymerization of styrene and methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Bromobenzeneacetic acid ethyl ester is used as an alkyl halide initiator in controlled/living radical polymerization of styrene and methyl methacrylate.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2882-19-1
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.10
MDL Number MFCD00013536
InChI Key BKTKLDMYHTUESO-UHFFFAOYNA-N
Synonym ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate
PubChem CID 97780
IUPAC Name ethyl 2-bromo-2-phenylacetate
SMILES CCOC(=O)C(Br)C1=CC=CC=C1

Specifications

Density 1.389
Boiling Point 164°C (30 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5395
Quantity 10 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 243.1
Percent Purity 97%
Chemical Name or Material Ethyl alpha-bromophenylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.