missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzimidate hydrochloride, 97%

Product Code. 11368626
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368626 10 g 10g
11378626 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368626 Supplier Thermo Scientific Alfa Aesar Supplier No. L17684.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5333-86-8 | C9H12ClNO | 185.65 g/mol

Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 5333-86-8
Molecular Formula C9H12ClNO
Molecular Weight (g/mol) 185.65
MDL Number MFCD00043246
InChI Key MODZVIMSNXSQIH-UHFFFAOYSA-N
Synonym ethyl benzimidate hydrochloride, benzenecarboximidic acid, ethyl ester, hydrochloride, ethyl benzenecarboximidate hydrochloride, benzimidic acid ethyl ester hcl, ethylbenzimidate hydrochloride, ethyl benzimidate hcl, ethyl benzylimidate hydrochloride, ethyl iminobenzoate hydrochloride, benzimido ethyl ether hydrochloride, benzimidoyl ethyl ether hydrochloride
PubChem CID 79249
IUPAC Name ethyl benzenecarboximidate;hydrochloride
SMILES [H+].[Cl-].CCOC(=N)C1=CC=CC=C1

Spezifikation

Melting Point 115°C to 117°C
Quantity 10 g
Beilstein 3697589
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 185.65
Percent Purity 97%
Chemical Name or Material Ethyl benzimidate hydrochloride

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.