missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzimidate hydrochloride, 97%

Product Code. 11378626
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378626 50 g 50g
11368626 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378626 Supplier Thermo Scientific Alfa Aesar Supplier No. L17684.18
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5333-86-8 | C9H12ClNO | 185.65 g/mol

Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzimidate hydrochloride is used in the preparation of (4R)-ethyl 2-phenyl-4,5-dihydrothiazole-4-carboxylate by reacting with (R)-ethyl cysteine hydrochloride. It also reacts with D-penicillamine methyl ester hydrochloride and triethylamine to prepare methyl-5,5- dimethyl-2-phenyl-2-thiazoline-4-carboxylate. Further, it is used as an intermediate for organic synthesis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5333-86-8
Molecular Formula C9H12ClNO
Molecular Weight (g/mol) 185.65
MDL Number MFCD00043246
InChI Key MODZVIMSNXSQIH-UHFFFAOYSA-N
Synonym ethyl benzimidate hydrochloride, benzenecarboximidic acid, ethyl ester, hydrochloride, ethyl benzenecarboximidate hydrochloride, benzimidic acid ethyl ester hcl, ethylbenzimidate hydrochloride, ethyl benzimidate hcl, ethyl benzylimidate hydrochloride, ethyl iminobenzoate hydrochloride, benzimido ethyl ether hydrochloride, benzimidoyl ethyl ether hydrochloride
PubChem CID 79249
IUPAC Name ethyl benzenecarboximidate;hydrochloride
SMILES [H+].[Cl-].CCOC(=N)C1=CC=CC=C1

Specifications

Melting Point 115°C to 117°C
Quantity 50 g
Beilstein 3697589
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 185.65
Percent Purity 97%
Chemical Name or Material Ethyl benzimidate hydrochloride
compliance-icons
Product Identifier
  • Ethyl benzimidate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl benzimidate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.