missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Benzoate, TRC

Product Code. 30372333
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372333 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372333 Supplier Toronto Research Chemicals Supplier No. TRCE89976010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoic Acid Ethylester
CAS 93-89-0
Purity Grade Notes HPLC
Molecular Formula C9 H10 O2
Quantity 10 g
Synonym Benzoic acid ethyl ester, Benzoyl ethyl ether, Ethyl benzenecarboxylate, Ethyl benzoate, NSC 8884
InChI Formula InChI=1S/C9H10O2/c1-2-11-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3
SMILES CCOC(=O)c1ccccc1
IUPAC Name ethyl benzoate
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.