missing translation for 'onlineSavingsMsg'
Learn More

(Ethyl benzoate)tricarbonylchromium, 98%, Thermo Scientific™

Product Code. 11389818
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389818 1 g 1g
11379818 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Afficher la politique du retour.
Code produit. 11389818 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' 044283.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Hydrogenation catalyst

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 32874-26-3
Molecular Formula C12H10CrO5
Molecular Weight (g/mol) 286.203
MDL Number MFCD02683559
InChI Key SWSAJUVURPSGIG-UHFFFAOYSA-N
Synonym ethyl benzoate tricarbonylchromium, ethyl benzoate tricarbonylchromium 0, tris carbon monoxide ethyl benzoate chromium
PubChem CID 13222494
IUPAC Name carbon monoxide;chromium;ethyl benzoate
SMILES CCOC(=O)C1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Spécification

Melting Point 54°C to 59°C
Color Orange
Quantity 1 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Formula Weight 286.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (Ethyl benzoate)tricarbonylchromium
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.