missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Benzoylformate, TRC

Product Code. 30381256
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381256 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381256 Supplier Toronto Research Chemicals Supplier No. TRCE8999955G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenyl-2-oxoacetic Acid Ethyl Ester
CAS 1603-79-8
Molecular Formula C10 H10 O3
Quantity 5 g
Synonym Glyoxylic acid, phenyl-, ethyl ester (6CI,7CI,8CI), 2-Oxo-2-phenylacetic acid ethyl ester, 2-Phenyl-2-oxoacetic acid ethyl ester, Ethyl 2-oxo-2-phenylacetate, Ethyl 2-phenyl-2-oxoacetate, Ethyl benzoylformate, Ethyl oxophenylacetate, Ethyl phenylglyoxylate, Ethyl α-oxobenzeneacetate, NSC 6766, Phenylglyoxylic acid ethyl ester, α-Oxobenzeneacetic acid ethyl ester
InChI Formula InChI=1S/C10H10O3/c1-2-13-10(12)9(11)8-6-4-3-5-7-8/h3-7H,2H2,1H3
SMILES CCOC(=O)C(=O)c1ccccc1
IUPAC Name ethyl 2-oxo-2-phenylacetate
Molecular Weight (g/mol) 178.18
Formula Weight 178.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.