missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Beta-chloropropionate, TRC

Product Code. 30375031
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375031 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375031 Supplier Toronto Research Chemicals Supplier No. TRCE67882525G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl beta-chloropropionate
CAS 623-71-2
Molecular Formula C5H9ClO2
Quantity 25 g
Synonym 3-Chloropropionic acid ethyl ester, Ethyl 3-chloropropanoateEthyl 3-chloropropionate, NSC 8845
InChI Formula InChI=1S/C5H9ClO2/c1-2-8-5(7)3-4-6/h2-4H2,1H3
SMILES CCOC(=O)CCCl
IUPAC Name ethyl 3-chloropropanoate
Molecular Weight (g/mol) 136.58
Formula Weight 136.0291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.