missing translation for 'onlineSavingsMsg'
Learn More

Ethyl beta-D-Thiogalactopyranoside, TRC

Product Code. 30376558
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376558 Supplier Toronto Research Chemicals Supplier No. TRCE9267751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl beta-D-Thiogalactopyranoside
CAS 56245-60-4
Molecular Formula C8 H16 O5 S
Quantity 1 g
InChI Formula InChI=1S/C8H16O5S/c1-2-14-8-7(12)6(11)5(10)4(3-9)13-8/h4-12H,2-3H2,1H3/t4-,5+,6+,7-,8+/m1/s1
SMILES CCS[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
IUPAC Name (2S,3R,4S,5R,6R)-2-ethylsulfanyl-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 224.27
Formula Weight 224.0718
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.