missing translation for 'onlineSavingsMsg'
Learn More

Ethyl caprate, 99+%

Product Code. 10377340 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
5mL
This item is not returnable. View return policy

Product Code. 10377340

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-38-3 | C12H24O2 | 200.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-38-3
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00009581
InChI Key RGXWDWUGBIJHDO-UHFFFAOYSA-N
Synonym ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
PubChem CID 8048
ChEBI CHEBI:87430
IUPAC Name ethyl decanoate
SMILES CCCCCCCCCC(=O)OCC

Specifications

Acidity 1mg KOH/g max.
Melting Point -20.0°C
Color Colorless
Density 0.8665g/mL
Boiling Point 245.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)8CO2C2H5
Refractive Index 1.4240 to 1.4270
Quantity 100 mL
Beilstein 02,356
Merck Index 15,3831
Specific Gravity 0.8665
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Viscosity <30 mPa.s (20°C)
Formula Weight 200.32
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl caprate, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.