missing translation for 'onlineSavingsMsg'
Learn More

Ethyl carbazate, 97%

Product Code. 10478450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10478450 25 g Glass bottle 25g
10122530 100 g Glass bottle 100g
10474925 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10478450 Supplier Thermo Scientific Acros Supplier No. 118200250
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 4114-31-2 | C3H8N2O2 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4114-31-2
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00007595
InChI Key VYSYZMNJHYOXGN-UHFFFAOYSA-N
Synonym ethyl carbazate, ethyl hydrazinecarboxylate, ethoxycarbohydrazide, hydrazinecarboxylic acid, ethyl ester, carbethoxyhydrazine, ethylcarbazate, ethyl carbazinate, carboethoxyhydrazine, monocarbethoxyhydrazine, n-carbethoxy hydrazine
PubChem CID 20064
IUPAC Name ethyl N-aminocarbamate
SMILES CCOC(=O)NN

Specifications

Melting Point 44°C to 48°C
Color Cream to White
Density 86g/mL
Boiling Point 108°C to 110°C (22.0 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NNHCO2C2H5
Quantity 25 g
Beilstein 03, 98
Fieser 01,360
Formula Weight 104.11
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Ethyl carbazate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl carbazate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.