missing translation for 'onlineSavingsMsg'
Learn More

Ethyl chloroacetate, 99%

Product Code. 10733464 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10733464

Brand: Thermo Scientific Acros 118222500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-39-5 | C4H7ClO2 | 122.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-39-5
MDL Number MFCD00000932
InChI Key VEUUMBGHMNQHGO-UHFFFAOYSA-N
Synonym ethyl chloroacetate, ethyl chloracetate, ethyl monochloroacetate, acetic acid, chloro-, ethyl ester, chloroacetic acid ethyl ester, ethyl monochloracetate, ethyl chloroethanoate, ethyl alpha-chloroacetate, 2-chloroacetic acid ethyl ester, unii-6h07teh4tf
PubChem CID 7751
IUPAC Name ethyl 2-chloroacetate
SMILES CCOC(=O)CCl

Specifications

Melting Point -26.0°C
Color Colorless
Density 1.1500g/mL
Boiling Point 142.0°C to 145.0°C
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7ClO2
Linear Formula ClCH2CO2C2H5
Refractive Index 1.4200 to 1.4220
Quantity 250 mL
Beilstein 02,197
Fieser 13,348
Merck Index 15,3838
Specific Gravity 1.15
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone
Molecular Weight (g/mol) 122.55
Viscosity 1.27 mPa.s (20°C)
Formula Weight 122.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl chloroacetate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl chloroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.