missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cinnamate, 98% trans

Product Code. 11994981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11994981 2.5 kg Glass bottle 2.5kg
10605012 100 g Glass bottle 100g
10615012 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11994981 Supplier Thermo Scientific Acros Supplier No. 150530025
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-36-6 | C11H12O2 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-36-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.21
InChI Key KBEBGUQPQBELIU-CMDGGOBGSA-N
Synonym ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
PubChem CID 637758
ChEBI CHEBI:4895
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
SMILES CCOC(=O)C=CC1=CC=CC=C1

Specifications

Acidity 1mg KOH/g max.
Melting Point 6.5°C to 7.5°C
Color Colorless to Yellow
Density 1.0460g/mL
Boiling Point 271.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCO2C2H5
Refractive Index 1.5580 to 1.5610
Quantity 2.5 kg
Merck Index 15,2299
Specific Gravity 1.046
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 176.21
Percent Purity 98% trans
Physical Form Liquid
Chemical Name or Material Ethyl cinnamate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.