missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cinnamate, Technical, Fisher Chemical

Product Code. 11428253 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Amber glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11428253 25 mL Amber glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11428253 Supplier Fisher Chemical Supplier No. E/1250/04
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 103-36-6
Melting Point 6.5°C
Color Yellow
Boiling Point 271°C
Packaging Amber glass bottle
Molecular Formula C11H12O2
MDL Number 9189
Quantity 25 mL
Synonym ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
InChI Key KBEBGUQPQBELIU-CMDGGOBGSA-N
SMILES CCOC(=O)C=CC1=CC=CC=C1
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
Molecular Weight (g/mol) 176.215
PubChem CID 637758
ChEBI CHEBI:4895
Formula Weight 176.21g/mol
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.