missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cinnamate, TRC

Product Code. 30387307
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387307 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387307 Supplier Toronto Research Chemicals Supplier No. TRCE903960100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinnamic acid ethylester
CAS 103-36-6
Molecular Formula C11 H12 O2
Quantity 100 g
Synonym Ethyl Benzylidene Acetate, 2-Propenoic acid, 3-phenyl-, ethyl ester, Cinnamic acid, ethyl ester (6CI,7CI,8CI), 3-Phenyl-2-propenoic acid ethyl ester, Ethyl 3-phenyl-2-propenoate, Ethyl 3-phenylacrylate, Ethyl benzylideneacetate, Ethyl cinnamate, NSC 6773, SemaSORB 9827
InChI Formula InChI=1S/C11H12O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-9H,2H2,1H3/b9-8+
SMILES CCOC(=O)\C=C\c1ccccc1
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
Molecular Weight (g/mol) 176.21
Formula Weight 176.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.