missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cyano(ethoxymethylene)acetate, TRC

Product Code. 30372335
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372335 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372335 Supplier Toronto Research Chemicals Supplier No. TRCE907510100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Cyano(ethoxymethylene)acetate
CAS 94-05-3
Molecular Formula C8H11NO3
Quantity 100 g
Synonym 2-(Ethoxymethylene)-2-cyanoacetic Acid Ethyl Ester, 2-Cyano-3-ethoxy-2-propenoic Acid Ethyl Ester, 2-Cyano-3-ethoxyacrylic Acid Ethyl Ester, Ethyl (ethoxymethylene)cyanoacetate, Ethyl 2-(Ethoxymethylene)-2-cyanoacetate, Ethyl 2-Cyano-2-(ethoxymethylene)acetate, Ethyl 2-Cyano-3-(ethyloxy)-2-propenoate, Ethyl 2-Cyano-3-ethoxy-2-propenoate, Ethyl 2-cyano-3-ethoxyacrylate, Ethyl 2-Cyano-3-ethoxypropenoate, Ethyl 2-ethoxymethylidenecyanoacetate, Ethyl 3-Ethoxy-2-cyanoacrylate, Ethyl Cyano(ethoxymethylene)acetate, Ethyl Cyano(ethoxymethylidene)acetate, Ethyl alpha-(Ethoxymethylene)-alpha-cyanoacetate, NSC 27797, NSC 31579, NSC 62026
InChI Formula InChI=1S/C8H11NO3/c1-3-11-6-7(5-9)8(10)12-4-2/h6H,3-4H2,1-2H3
SMILES CCOC=C(C#N)C(=O)OCC
IUPAC Name ethyl 2-cyano-3-ethoxyprop-2-enoate
Molecular Weight (g/mol) 169.18
Formula Weight 169.0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.