missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Cyano(hydroxyimino)acetate, TRC

Product Code. 30387304
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387304 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387304 Supplier Toronto Research Chemicals Supplier No. TRCE918840100G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Cyano(hydroxyimino)acetate
CAS 3849-21-6
Molecular Formula C5H6N2O3
Quantity 100 g
Synonym Cyano(hydroxyimino)-acetic Acid Ethyl Ester, 2-Cyano-2-(hydroxyimino)acetic Acid Ethyl Ester, Ethyl 2-(Hydroxyimino)cyanoacetate, Ethyl 2-Cyano-2-(hydroxyimino)acetate, Ethyl 2-Cyano-2-(hydroxyimino)ethanoate, Ethyl 2-Cyano-2-oximinoacetate, Ethyl 2-Hydroxyimino-2-cyanoacetate, Ethyl Cyano(hydroxyimino)acetate, Ethyl Cyanoglyoxylate Oxime, Ethyl Cyanoglyoxylate-2-oxime, Ethyl alpha-Cyano-alpha-oximinoacetate, NSC 137667, NSC 41021, NSC 42116, Oxyma Pure, (E)-Ethyl 2-Cyano-2-(hydroxyimino)acetate
InChI Formula InChI=1S/C5H6N2O3/c1-2-10-5(8)4(3-6)7-9/h9H,2H2,1H3/b7-4+
SMILES CCOC(=O)C(=NO)C#N
Molecular Weight (g/mol) 142.11
Formula Weight 142.0378
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.