missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyanoacetate, 98+%

Product Code. 11329621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11329621 5 kg Plastic drum 5kg
10617652 250 g Glass bottle 250g
10102630 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329621 Supplier Thermo Scientific Acros Supplier No. 118280051
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-56-6 | C5H7NO2 | 113.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-56-6
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00001940
InChI Key ZIUSEGSNTOUIPT-UHFFFAOYSA-N
Synonym ethyl cyanoacetate, ethyl cyanacetate, cyanoacetic acid ethyl ester, acetic acid, cyano-, ethyl ester, ethyl cyanoethanoate, cyanoacetic ester, cyanacetate ethyle, usaf kf-25, estere cianoacetico, ethyl cyano acetate
PubChem CID 7764
IUPAC Name ethyl 2-cyanoacetate
SMILES CCOC(=O)CC#N

Specifications

Melting Point -22.0°C
Density 1.0600g/mL
Boiling Point 208.0°C to 210.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic drum
Linear Formula NCCH2CO2C2H5
Refractive Index 1.4170 to 1.4190
Quantity 5 kg
Beilstein 02,585
Fieser 13,69
Merck Index 15,3841
Specific Gravity 1.06
Solubility Information Solubility in water: 20g/L(20°C). Other solubilities: soluble in ammonium hydroxide,diethyl ether,,ethanol,aqueous solutions of alkalies
Viscosity 1.01 mPa.s (20°C)
Formula Weight 113.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Ethyl cyanoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl cyanoacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.