missing translation for 'onlineSavingsMsg'
Learn More

Ethyl cyclohexylideneacetate, 98%

Product Code. 11496057
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496057 100 g 100g
11476057 5 g 5g
11486057 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496057 Supplier Thermo Scientific Alfa Aesar Supplier No. B25073.22
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1552-92-7 | C10H16O2 | 168.24 g/mol

Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyclohexylideneacetate is used as pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl acetate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1552-92-7
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00017627,MFCD00963857
InChI Key MCWDXHYYYNGYGK-UHFFFAOYSA-N
Synonym ethyl cyclohexylideneacetate, acetic acid, cyclohexylidene-, ethyl ester, ethyl 2-cyclohexylidenacetate, cyclohexylideneacetic acid ethyl ester, ethoxycarbonylmethylenecyclohexane, .delta.1,.alpha.-cyclohexaneacetic acid, ethyl ester, acmc-20ahxr, ethylcyclohexylidenacetate, .delta.1, ethyl ester, ethyl2-cyclohexylideneacetate
PubChem CID 73776
IUPAC Name ethyl 2-cyclohexylideneacetate
SMILES CCOC(=O)C=C1CCCCC1

Specifications

Boiling Point 48°C to 49°C (0.02 mmHg)
Quantity 100 g
Solubility Information Soluble in dichloromethane,ethyl acetate.
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Ethyl cyclohexylideneacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.