missing translation for 'onlineSavingsMsg'
Learn More

Ethyl dibromoacetate, 96%

Product Code. 11470807
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470807 5 g 5g
11480807 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470807 Supplier Thermo Scientific Alfa Aesar Supplier No. B23209.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 617-33-4 | C4H6Br2O2 | 245.898 g/mol

Ethyl dibromoacetate can be used to produce diphenoxy-acetic acid ethyl ester. It is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl dibromoacetate can be used to produce diphenoxy-acetic acid ethyl ester. It is used in organic synthesis.

Solubility
Soluble in hexane, ether, CH2Cl2, acetone, EtOAc, ethanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-33-4
Molecular Formula C4H6Br2O2
Molecular Weight (g/mol) 245.898
MDL Number MFCD00041718
InChI Key NIJGVVHCUXNSLL-UHFFFAOYSA-N
Synonym ethyl dibromoacetate, acetic acid, dibromo-, ethyl ester, dibromoacetic acid, ethyl ester, acetic acid, 2,2-dibromo-, ethyl ester, ethyldibromoacetate, ethyl dibromo acetate, acmc-20akxm, aceticacid,dibromo-,ethylester, dibromoacetic acid ethyl ester, dibromo-acetic acid ethyl ester
PubChem CID 69237
IUPAC Name ethyl 2,2-dibromoacetate
SMILES CCOC(=O)C(Br)Br

Specifications

Density 1.902
Boiling Point 76°C to 78°C (12 mmHg)
Linear Formula Br2CHCO2CH2CH3
Refractive Index 1.499
Quantity 5 g
Solubility Information Soluble in hexane,ether,CH2Cl2,acetone,EtOAc,ethanol.
Formula Weight 245.9
Percent Purity 96%
Chemical Name or Material Ethyl dibromoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.